Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wasf1
Entrez Gene ID
294568
Gene Name
WAS protein family, member 1
Gene Aliases
-
Chromosome Location
Chr.20: 48456191 - 48503768 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025114.1 NP_001020285.1 4 - 5 490 67
XM_006256586.2 XP_006256648.1 4 - 5 683 67
XM_006256587.2 XP_006256649.1 4 - 5 684 67
GenBank mRNA BC091322.1 - 4 - 5 490 67

View Full Product Details