The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024781.1 | NP_001019952.1 | 1 - 2 | 444 | 55 |
GenBank mRNA | BC097973.1 | - | 1 - 2 | 444 | 55 |