Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lsm4
Entrez Gene ID
290647
Gene Name
LSM4 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.16: 20506399 - 20511818 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106073.1 NP_001099543.1 4 - 5 520 120

View Full Product Details