Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ids
Entrez Gene ID
363513
Gene Name
iduronate 2-sulfatase
Gene Aliases
-
Chromosome Location
Chr.8: 69447971 - 69466708 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008766428.1 XP_008764650.1 8 - 9 1432 66

View Full Product Details