Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nip30
Entrez Gene ID
307652
Gene Name
NEFA-interacting nuclear protein NIP30
Gene Aliases
RGD1307433
Chromosome Location
Chr.19: 10827057 - 10856747 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014014.1 NP_001014036.1 4 - 5 483 77
XM_006255110.2 XP_006255172.1 4 - 5 793 77
XM_006255111.2 XP_006255173.1 4 - 5 560 77
XM_006255114.2 XP_006255176.1 5 - 6 1025 77
GenBank mRNA BC079145.1 - 4 - 5 483 77

View Full Product Details