Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ogfod1
Entrez Gene ID
307657
Gene Name
2-oxoglutarate and iron-dependent oxygenase domain containing 1
Gene Aliases
RGD1308848
Chromosome Location
Chr.19: 11425301 - 11451085 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107411.2 NP_001100881.1 4 - 5 448 106

View Full Product Details