Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Olr75
Entrez Gene ID
365327
Gene Name
olfactory receptor 75
Gene Aliases
-
Chromosome Location
Chr.1: 168204985 - 168205944 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001281.1 NP_001001281.1 1 - 1 459 123

View Full Product Details