Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajb13
Entrez Gene ID
308857
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 13
Gene Aliases
Tsarg1
Chromosome Location
Chr.1: 165515718 - 165530163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005885.2 NP_001005885.1 3 - 4 338 154
XM_006229766.2 XP_006229828.1 3 - 4 403 154
GenBank mRNA AY325765.1 - 3 - 4 378 154
AY380804.1 - 3 - 4 396 154
BC098002.1 - 3 - 4 338 154

View Full Product Details