Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajb13
Entrez Gene ID
308857
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 13
Gene Aliases
Tsarg1
Chromosome Location
Chr.1: 165515718 - 165530163 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005885.2 NP_001005885.1 6 - 7 725 68
XM_006229766.2 XP_006229828.1 6 - 7 790 68
GenBank mRNA AY325765.1 - 6 - 7 765 68
AY380804.1 - 6 - 7 783 68
BC098002.1 - 6 - 7 725 68

View Full Product Details