Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsmce1
Entrez Gene ID
361645
Gene Name
non-SMC element 1 homolog (S. cerevisiae)
Gene Aliases
RGD1307760
Chromosome Location
Chr.1: 196855521 - 196883908 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039611.1 NP_001034700.1 3 - 4 398 73
XM_006230291.2 XP_006230353.1 2 - 3 240 73
XM_006230292.2 XP_006230354.1 2 - 3 810 73
XM_006230293.2 XP_006230355.1 2 - 3 320 73
GenBank mRNA BC099757.1 - 3 - 4 398 73

View Full Product Details