Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Abca15
Entrez Gene ID
293442
Gene Name
ATP-binding cassette, subfamily A (ABC1), member 15
Gene Aliases
-
Chromosome Location
Chr.1: 190072420 - 190178813 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106293.2 NP_001099763.2 22 - 23 3761 72

View Full Product Details