Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbp1
Entrez Gene ID
171091
Gene Name
Z-DNA binding protein 1
Gene Aliases
Dlm1
Chromosome Location
Chr.3: 171276062 - 171294844 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133564.1 NP_598248.1 3 - 4 468 95
XM_006235674.2 XP_006235736.1 4 - 5 4311 95
GenBank mRNA AJ302054.1 - 3 - 4 468 95

View Full Product Details