Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grm3
Entrez Gene ID
24416
Gene Name
glutamate receptor, metabotropic 3
Gene Aliases
mGluR3
Chromosome Location
Chr.4: 21317219 - 21562592 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105712.1 NP_001099182.1 1 - 2 83 76
XM_008762682.1 XP_008760904.1 1 - 2 559 76

View Full Product Details