The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001106206.1 | NP_001099676.1 | 4 - 5 | 1131 | 70 |
GenBank mRNA | BC158745.1 | - | 4 - 5 | 1071 | 70 |