The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013910.2 | NP_001013932.1 | 2 - 3 | 168 | 61 |
GenBank mRNA | BC097370.1 | - | 2 - 3 | 168 | 61 |