The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006232238.2 | XP_006232300.1 | 5 - 6 | 1699 | 60 |
XM_006232240.2 | XP_006232302.1 | 7 - 8 | 1428 | 60 | |
XM_008760950.1 | XP_008759172.1 | 8 - 9 | 3616 | 60 |