Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ctif
Entrez Gene ID
364900
Gene Name
CBP80/20-dependent translation initiation factor
Gene Aliases
Gm672
Chromosome Location
Chr.18: 71467381 - 71701423 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108891.1 NP_001102361.1 3 - 4 917 93

View Full Product Details