The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013228.1 | NP_001013246.1 | 3 - 4 | 226 | 94 |
GenBank mRNA | BC083797.1 | - | 3 - 4 | 226 | 94 |