Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Antxrl
Entrez Gene ID
364513
Gene Name
anthrax toxin receptor-like
Gene Aliases
RGD1563190
Chromosome Location
Chr.16: 10383973 - 10407610 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252719.2 XP_006252781.2 1 - 2 278 117

View Full Product Details