Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam20c
Entrez Gene ID
304334
Gene Name
family with sequence similarity 20, member C
Gene Aliases
RGD1311980
Chromosome Location
Chr.12: 17913771 - 17972733 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012238.1 NP_001012238.1 8 - 9 1957 88
GenBank mRNA BC161825.1 - 8 - 9 1624 88

View Full Product Details