Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdk8
Entrez Gene ID
498140
Gene Name
cyclin-dependent kinase 8
Gene Aliases
RGD1560888
Chromosome Location
Chr.12: 10496273 - 10561335 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109061.1 NP_001102531.1 6 - 7 661 78

View Full Product Details