Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Grid2ip
Entrez Gene ID
288484
Gene Name
glutamate receptor, ionotropic, delta 2 (Grid2) interacting protein
Gene Aliases
-
Chromosome Location
Chr.12: 13221372 - 13257489 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105910.1 NP_001099380.1 3 - 4 404 62
XM_006248865.2 XP_006248927.1 3 - 4 474 62
XM_006248866.2 XP_006248928.1 4 - 5 1066 62

View Full Product Details