Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ing1
Entrez Gene ID
306626
Gene Name
inhibitor of growth family, member 1
Gene Aliases
p33ING1b, p33ING1c
Chromosome Location
Chr.16: 83277055 - 83278120 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY624103.1 - - 93 79
AY624104.1 - - 93 79

View Full Product Details