Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Proz
Entrez Gene ID
306608
Gene Name
protein Z, vitamin K-dependent plasma glycoprotein
Gene Aliases
-
Chromosome Location
Chr.16: 81784348 - 81797901 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253473.1 XP_006253535.1 3 - 4 380 56
GenBank mRNA BC160868.1 - 3 - 4 292 56
FQ209855.1 - 3 - 4 294 56

View Full Product Details