Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shcbp1
Entrez Gene ID
364648
Gene Name
Shc SH2-domain binding protein 1
Gene Aliases
-
Chromosome Location
Chr.16: 90613825 - 90642683 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008771421.1 XP_008769643.1 7 - 8 1132 77

View Full Product Details