Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Upf3a
Entrez Gene ID
361176
Gene Name
UPF3 regulator of nonsense transcripts homolog A (yeast)
Gene Aliases
-
Chromosome Location
Chr.16: 81521084 - 81532512 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012159.1 NP_001012159.1 7 - 8 870 196
GenBank mRNA BC088463.1 - 7 - 8 870 196

View Full Product Details