Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tcte1
Entrez Gene ID
316242
Gene Name
t-complex-associated testis expressed 1
Gene Aliases
-
Chromosome Location
Chr.9: 17847871 - 17867283 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108206.1 NP_001101676.1 1 - 2 342 61
XM_006244539.2 XP_006244601.1 2 - 3 865 61
XM_006244540.2 XP_006244602.1 3 - 4 942 61

View Full Product Details