Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpl7l1
Entrez Gene ID
317275
Gene Name
ribosomal protein L7-like 1
Gene Aliases
RGD1308469
Chromosome Location
Chr.9: 16508477 - 16516502 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_228615.8 XP_228615.3 2 - 3 440 92
GenBank mRNA FQ214078.1 - 2 - 3 142 92

View Full Product Details