Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pla2g7
Entrez Gene ID
301265
Gene Name
phospholipase A2, group VII (platelet-activating factor acetylhydrolase, plasma)
Gene Aliases
-
Chromosome Location
Chr.9: 19935754 - 19978013 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009353.1 NP_001009353.1 9 - 10 1091 83
XM_006244606.2 XP_006244668.1 9 - 10 1208 83
XM_008766856.1 XP_008765078.1 8 - 9 2851 83
GenBank mRNA BC088457.1 - 9 - 10 1091 83

View Full Product Details