Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ripk1
Entrez Gene ID
306886
Gene Name
receptor (TNFRSF)-interacting serine-threonine kinase 1
Gene Aliases
-
Chromosome Location
Chr.17: 31537591 - 31569904 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107350.1 NP_001100820.1 2 - 3 518 127

View Full Product Details