Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rpp40
Entrez Gene ID
291071
Gene Name
ribonuclease P/MRP 40 subunit
Gene Aliases
-
Chromosome Location
Chr.17: 29638663 - 29647709 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253790.1 XP_006253852.1 1 - 2 146 82

View Full Product Details