Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Acer1
Entrez Gene ID
301118
Gene Name
alkaline ceramidase 1
Gene Aliases
Asah3
Chromosome Location
Chr.9: 10061978 - 10093588 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106875.1 NP_001100345.1 2 - 3 256 96

View Full Product Details