The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107815.2 | NP_001101285.1 | 5 - 6 | 650 | 63 |
XM_006233594.2 | XP_006233656.1 | 6 - 7 | 710 | 63 | |
XM_006233596.2 | XP_006233658.1 | 5 - 6 | 554 | 63 | |
XM_008761588.1 | XP_008759810.1 | 6 - 7 | 656 | 63 | |
GenBank mRNA | BC169064.1 | - | 5 - 6 | 650 | 63 |