The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033868.1 | NP_001029040.1 | 5 - 6 | 667 | 118 |
GenBank mRNA | CK358924.1 | - | 5 - 6 | 601 | 118 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006233722.2 | XP_006233784.1 | 4 - 5 | 2519 | 118 |