Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Blzf1
Entrez Gene ID
498266
Gene Name
basic leucine zipper nuclear factor 1
Gene Aliases
-
Chromosome Location
Chr.13: 82592170 - 82607379 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017494.1 NP_001017494.1 5 - 6 954 101
GenBank mRNA BC079114.1 - 5 - 6 954 101

View Full Product Details