Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ncf2
Entrez Gene ID
364018
Gene Name
neutrophil cytosolic factor 2
Gene Aliases
-
Chromosome Location
Chr.13: 70226434 - 70259019 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100984.1 NP_001094454.1 9 - 10 828 61
XM_006250013.2 XP_006250075.1 8 - 9 1047 61
XM_008769664.1 XP_008767886.1 6 - 7 819 61
GenBank mRNA JN864041.1 - 7 - 8 852 61
JN864042.1 - 7 - 8 852 61
JN864045.1 - 7 - 8 846 61
JN864046.1 - 6 - 7 738 61

View Full Product Details