Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lmx1a
Entrez Gene ID
289201
Gene Name
LIM homeobox transcription factor 1 alpha
Gene Aliases
-
Chromosome Location
Chr.13: 85917956 - 86061536 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105967.2 NP_001099437.2 2 - 3 483 79
XM_008769728.1 XP_008767950.1 2 - 3 376 79

View Full Product Details