Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rabif
Entrez Gene ID
304807
Gene Name
RAB interacting factor
Gene Aliases
Mss4
Chromosome Location
Chr.13: 51317476 - 51330536 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007678.2 NP_001007679.1 1 - 2 145 61
GenBank mRNA BC161807.1 - 1 - 2 153 61
CF977877.1 - 1 - 2 169 61

Gene Symbol
RGD1563962
Entrez Gene ID
499636
Gene Name
similar to Mss4 protein
Gene Aliases
-
Chromosome Location
Chr.2: 165805230 - 165816503 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003749301.3 XP_003749349.1 1 - 2 139 61

View Full Product Details