Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fech
Entrez Gene ID
361338
Gene Name
ferrochelatase
Gene Aliases
-
Chromosome Location
Chr.18: 59941992 - 59975192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108434.1 NP_001101904.1 1 - 2 165 66

View Full Product Details