Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdc14b
Entrez Gene ID
361195
Gene Name
cell division cycle 14B
Gene Aliases
-
Chromosome Location
Chr.17: 1709703 - 1797732 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108404.1 NP_001101874.1 3 - 4 507 95

View Full Product Details