Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdca5
Entrez Gene ID
684771
Gene Name
cell division cycle associated 5
Gene Aliases
RGD1560863
Chromosome Location
Chr.1: 221448661 - 221462235 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760176.1 XP_008758398.1 5 - 6 775 76

View Full Product Details