Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cdc42ep2
Entrez Gene ID
309175
Gene Name
CDC42 effector protein (Rho GTPase binding) 2
Gene Aliases
-
Chromosome Location
Chr.1: 221272349 - 221281246 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006230806.2 XP_006230868.1 1 - 2 89 61

View Full Product Details