Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il22
Entrez Gene ID
500836
Gene Name
interleukin 22
Gene Aliases
RGD1561292
Chromosome Location
Chr.7: 61236037 - 61240502 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191988.1 NP_001178917.1 3 - 4 396 78
GenBank mRNA DQ630444.1 - 3 - 4 375 78

View Full Product Details