Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsp90b1
Entrez Gene ID
362862
Gene Name
heat shock protein 90, beta, member 1
Gene Aliases
Grp94, Tra1
Chromosome Location
Chr.7: 27226570 - 27240533 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012197.2 NP_001012197.2 3 - 4 420 150
GenBank mRNA BC081917.1 - 3 - 4 420 150

View Full Product Details