Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mapre2
Entrez Gene ID
679221
Gene Name
microtubule-associated protein, RP/EB family, member 2
Gene Aliases
-
Chromosome Location
Chr.18: 14814150 - 15780290 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101000.1 NP_001094470.1 - 781 93
GenBank mRNA BC105879.1 - - 781 93

View Full Product Details