The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001191079.1 | NP_001178008.1 | 6 - 7 | 684 | 63 |
XM_008771934.1 | XP_008770156.1 | 6 - 7 | 684 | 63 |