Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tslp
Entrez Gene ID
688621
Gene Name
thymic stromal lymphopoietin
Gene Aliases
-
Chromosome Location
Chr.18: 25613655 - 25618066 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008772052.1 XP_008770274.1 3 - 4 188 85

View Full Product Details