Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Commd2
Entrez Gene ID
688478
Gene Name
COMM domain containing 2
Gene Aliases
-
Chromosome Location
Chr.2: 147774767 - 147779013 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109503.1 NP_001102973.1 4 - 5 459 70
GenBank mRNA BC098743.1 - 3 - 4 1028 70
FQ229674.1 - 4 - 5 403 70

View Full Product Details