Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccna1
Entrez Gene ID
295052
Gene Name
cyclin A1
Gene Aliases
-
Chromosome Location
Chr.2: 144456689 - 144469255 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011949.1 NP_001011949.1 7 - 8 1355 102
XM_006232359.2 XP_006232421.1 7 - 8 1317 102
XM_006232360.2 XP_006232422.1 7 - 8 1320 102
XM_006232361.2 XP_006232423.1 7 - 8 1360 102
XM_006232363.2 XP_006232425.1 7 - 8 1248 102
XM_008760987.1 XP_008759209.1 7 - 8 1410 102
XM_008760988.1 XP_008759210.1 7 - 8 1301 102
GenBank mRNA BC079234.1 - 7 - 8 1355 102

View Full Product Details