Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P2ry14
Entrez Gene ID
171108
Gene Name
purinergic receptor P2Y, G-protein coupled, 14
Gene Aliases
Gpr105
Chromosome Location
Chr.2: 149331948 - 149379336 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008760984.1 XP_008759206.1 1 - 2 5320 68
GenBank mRNA BC083545.1 - 1 - 2 79 68

View Full Product Details